Learn More
Sino Biological Mouse LIF Protein 1MG (Sino Biological)
Supplier:  Sino Biological 50755M08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse LIF (P09056) (Met 1-Phe 203) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3466