Learn More
Sino Biological Recombinant Anti-LIFR Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50423R110100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LIFR (rM LIFR; Catalog#50423-M08H; NP_038612.1; Met 1-Ser 828).
Catalog No. 50-241-2067