Learn More
Sino Biological Anti-LIFR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50423T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse LIFR (rM LIFR; Catalog#50423-M08H; NP_038612.1; Met1-Ser828). LIFR specific IgG was purified by Mouse LIFR affinity chromatography.
Catalog No. 50-241-1458