Learn More
Sino Biological Anti-LIFR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50423RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse LIFR (rM LIFR; Catalog#50423-M08H; NP_038612.1; Met 1-Ser 828). LIFR specific IgG was purified by mouse LIFR affinity chromatography.
Catalog No. 50-240-9968