Learn More
Sino Biological Anti-ILT5/LILRB3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50760T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse LILRB3 (rM LILRB3; Catalog#50760-M08H; AAC53219.1; Met1-Tyr640). LILRB3 specific IgG was purified by Mouse LILRB3 affinity chromatography.
Catalog No. 50-241-0926