Learn More
Alpco Ltd MOUSE LIPOCALIN-2/NGAL ELISA
SDP
Supplier:  Alpco Ltd 41NGALMSE01
Encompass
Mouse Lipocalin-2/NGAL ELISA is for the quantitative determination of Lipocalin-2/NGAL in mouse plasma, serum, and urine samples. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2151667