Learn More
American Research Products Inc Mouse lipoprotein lipase LPL
Catalog No. 5000326133
Encompass
Mouse lipoprotein lipase LPL ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26133 Supplier American Research Products Inc Supplier No. CSBE08495M
Mouse lipoprotein lipase LPL ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich