Learn More
Cayman Chemical Mouse LIvR X RECPTR Alpha
SDP
Catalog No. 502909389
Encompass_Preferred
mLXRα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog No. M00311, M00311-32]
Catalog No. 50-290-9389 Supplier Cayman Chemical Supplier No. 367113X32 WELL
Add to Cart
Add to Cart
mLXRα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog No. M00311, M00311-32]