Learn More
Cayman Chemical Mouse Liver X Receptor Beta Reporter Assay System, 3x32 Well
SDP
Catalog No. 502909390
Encompass_Preferred
mLXRβ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. M00301, M00301-32]
Catalog No. 50-290-9390 Supplier Cayman Chemical Supplier No. 367123X32 WELL
mLXRβ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. M00301, M00301-32]