Learn More
BIOHIPPO Mouse LMNA Microsample
SDP
Catalog No. 5000314389
Encompass
Mouse LMNA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LMNA in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 113 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse LMNA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse LMNA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse LMNA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14389 Supplier BIOHIPPO Supplier No. BHE15211102
Add to Cart
Add to Cart
Mouse LMNA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LMNA in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 113 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse LMNA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse LMNA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse LMNA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color