Learn More
BIOHIPPO Mouse LN Microsample
SDP
Catalog No. 5000314774
Encompass
Mouse LN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LN in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 27 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse LN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse LN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse LN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14774 Supplier BIOHIPPO Supplier No. BHE15212660
Mouse LN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LN in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 27 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse LN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse LN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse LN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color