Learn More
Biomatik Corporation Mouse Low Density Lipoprotein Receptor Related Protein 2 (LRP2) ELISA Kit
Supplier:  Biomatik Corporation EKU12207
Encompass_Preferred
Mouse Low Density Lipoprotein Receptor Related Protein 2 (LRP2) ELISA Kit for detecting LRP2 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.123ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A2ARV4. Gene ID: 14725. Target Synonyms: DBS; gp330; Glycoprotein 330; Megalin; Low Density Lipoprotein-Related Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8006