Learn More
American Research Products Inc Mouse low density lipoprotein
Supplier:  American Research Products Inc CSBE13476M
Encompass
Mouse low density lipoprotein LDL ELISA Kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29255