Learn More
BIOHIPPO Mouse LpPLA2 Microsample
SDP
Catalog No. 5000315204
Encompass
Mouse LpPLA2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LpPLA2 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 0 132 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse LpPLA2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse LpPLA2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse LpPLA2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15204 Supplier BIOHIPPO Supplier No. BHE15212655
Mouse LpPLA2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LpPLA2 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 0 132 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse LpPLA2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse LpPLA2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse LpPLA2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color