Learn More
Sino Biological Mouse LRPAP1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 50281R009100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LRPAP1 (rM LRPAP1; Catalog#50281-M08H; NP_038615.2; Gln 29-Leu 360).
Catalog No. 50-237-1865