Learn More
Sino Biological Anti-LRPAP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50281RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse LRPAP1 (rM LRPAP1; Catalog#50281-M08H; NP_038615.2; Gln 29-Leu 360). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1291