Learn More
Sino Biological Recombinant Mouse LRRC15 Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 5A4998M49HB100
Encompass_Preferred
A DNA sequence encoding the Mouse LRRC15 (NP_083249.1) (Met1-Gly536) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-2441