Learn More
R&D Systems™ Mouse LRRC3 (NP_660134) VersaClone cDNA
Description
- Leucine-rich Repeat Containing 3
- Also known as: C21orf102; chromosome 21 open reading frame 102; leucine rich repeat containing 3; leucine-rich repeat-containing protein 3; LRRC3; LRRC3A
- The LRRC3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- A 25 kDa (predicted) member of the Leu-rich repeat protein superfamily.
- Little is known about the protein except for the fact that it shows widespread expression.
- Human LRRC3 is a protein that is 225 amino acids (aa) in length (SwissProt #:Q9BY71).
- It contains three LRRs between aa 63-86, aa 87-110, and aa 112-135.
- There is one potential splice variant that shows a deletion of aa 129-248.
- Over aa 33-204, human LRRC3 is 83% and 81% aa identical to mouse and canine LRRC3, respectively.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mLrrc3 |
| Accession Number | NP_660134 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 787 bp |
| Gene ID (Entrez) | 81543 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.