Learn More
Sino Biological Recombinant Mouse NGL-1/LRRC4C Protein (ECD, His Tag)
Supplier:  Sino Biological 50529M08H50
Encompass_Preferred
A DNA sequence encoding the mouse LRRC4C (BAC27884.1) (Met1-Lys527) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3222