Learn More
BIOHIPPO Mouse LTb Microsample
SDP
Catalog No. 5000314494
Encompass
Mouse LTb Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LTb in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse LTb Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse LTb Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse LTb biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14494 Supplier BIOHIPPO Supplier No. BHE15211572
Mouse LTb Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LTb in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse LTb Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse LTb Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse LTb biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color