Learn More
Sino Biological Mouse LXN Protein 50ug (Sino Biological)
Supplier:  Sino Biological 50050M07E50
Encompass_Preferred
A DNA sequence encoding the mouse LXN (NP_058033.2) (Glu 2-Glu 222) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-237-2547