Learn More
Sino Biological Recombinant Mouse Ly-6A/E (Sca-1) Protein (His Tag)
Supplier:  Sino Biological 53809M07E100
Encompass_Preferred
A DNA sequence encoding the mouse LY6A (NP_001258345.1) (Leu27-Gly119) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-2963