Learn More
ACROBiosystems Mouse LY75 / CD205 Protein, His Tag
Catalog No. 502054857
Encompass_Preferred
Recombinant Protein;1MG;Mouse LY75, His Tag (LY5-M52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Asp 1667 (Accession # Q60767-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 190.0 kDa.
Catalog No. 50-205-4857 Supplier ACROBiosystems Supplier No. LY5M52H51MG
Recombinant Protein;1MG;Mouse LY75, His Tag (LY5-M52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Asp 1667 (Accession # Q60767-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 190.0 kDa.