Learn More
Sino Biological Recombinant Human RAGE Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50062M08H1
Encompass_Preferred
A DNA sequence encoding the mature form of human AGER (NP_001127) extracellular domain (Met1-Ala 344) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7157