Learn More
Sino Biological Recombinant Anti-CD229/Ly9/SLAMF3 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50504R002P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Ly9 / CD229 / SLAMF3 (rM Ly9 / CD229 / SLAMF3; Catalog#50504-M08H; NP_032560.2; Met1-Arg453) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-3233