Learn More
Sino Biological Recombinant Anti-CD229/Ly9/SLAMF3 Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 50504R001H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LY9 / CD229 / SLAMF3 (rM LY9 / CD229 / SLAMF3; Catalog#50504-M08H; NP_032560.2; Met1-Arg453). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-3241