Learn More
Sino Biological Recombinant Anti-CD229/Ly9/SLAMF3 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50504R00120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD229 (rM CD229; Catalog#50504-M08H; NP_032560.2; Met1-Arg453).
Catalog No. 50-255-3242