Learn More
Sino Biological Anti-CD229/Ly9/SLAMF3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50504RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse LY9 / CD229 / SLAMF3 (rM LY9 / CD229 / SLAMF3; Catalog#50504-M08H; NP_032560.2; Met 1-Arg 453). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3229