Learn More
Biomatik Corporation Mouse Lymphocyte Activation Gene 3 (LAG3) ELISA Kit
Supplier:  Biomatik Corporation EKU09073
Encompass_Preferred
Mouse Lymphocyte Activation Gene 3 (LAG3) ELISA Kit for detecting LAG3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61790. Gene ID: 16768. Target Synonyms: CD223; Protein FDC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6243