Learn More
American Research Products Inc Mouse lymphotactin Lptn LTN E
Catalog No. 5000328707
Encompass
Mouse lymphotactin Lptn/LTN ELISA Kit from CUSABIO Detection Range 0 156 ng/ml - 10 ng/ml Sensitivity 0 039 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28707 Supplier American Research Products Inc Supplier No. CSBE08714M
Mouse lymphotactin Lptn/LTN ELISA Kit from CUSABIO Detection Range 0 156 ng/ml - 10 ng/ml Sensitivity 0 039 ng/ml Sample serum plasma tissue homogenates Method Sandwich