Learn More
Sino Biological Mouse Lymphotactin/XCL1/SCYC1 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786748
Encompass_Preferred
The Mouse XCL1 ELISA Pair Set is for the quantitative determination of Mouse XCL1.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse XCL1 coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse XCL1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse XCL1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse XCL1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86748 Supplier Sino Biological Supplier No. SEK5067715
The Mouse XCL1 ELISA Pair Set is for the quantitative determination of Mouse XCL1.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse XCL1 coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse XCL1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse XCL1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse XCL1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.