Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse LYPD1 (NP_659568) VersaClone cDNA

Catalog No. RDC1924
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC1924 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC1924 Supplier R&D Systems™ Supplier No. RDC1924
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Mouse LYPD1 (NP_659568) VersaClone cDNA is a member of the Ly6 family of GPI-anchored proteins.

The Mouse LYPD1 (NP_659568) VersaClone cDNA clone contains the complete ORF for LYPD1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The LYPD1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Specifications

Gene mLypd1
Accession Number NP_659568
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 439bp
pH 8.5
Gene ID (Entrez) 116372
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.