Learn More
American Research Products Inc Mouse Lysosome associated memb
Catalog No. 5000327103
Encompass
Mouse Lysosome-associated membrane glycoprotein 2 (LAMP2) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27103 Supplier American Research Products Inc Supplier No. CSBEL012740MO
Mouse Lysosome-associated membrane glycoprotein 2 (LAMP2) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich