Learn More
Kingfisher Biotech Inc Mouse M-CSF Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0462M025
Encompass_Preferred
The Mouse M-CSF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Mouse M-CSF applications are for cell culture, ELISA standard, and Western Blot Control. The Mouse M-CSF yeast-derived recombinant protein can be purchased in multiple sizes. Mouse M-CSF Specifications: (Molecular Weight: 26.0 kDa) (Amino Acid Sequence: AKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKPDCNC LYPKATPSSD PASASPHQPP APSMAPLAGL AWDDSQRTEG SSLLPSELPL RIEDPGSAKQ RPPRSTCQTL E (231)) (Gene ID: 12977). For research use only. Made in the USA
Catalog No. 50-241-3462