Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse M-CSF, Animal-Free Recombinant Protein, PeproTech®

Catalog No. 50398632
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 μg
250 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-398-632 50 μg
AF31502250U 250 μg
50-162-8688 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-398-632 Supplier Gibco™ Supplier No. AF3150250UG
  • $656.00
    $557.65 / Each of 1

    Save $98.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Recombinant Murine M-CSF is a 36.4 kDa homodimeric protein consisting of two 156 amino acid polypeptide subunits. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

M-CSF (Macrophage colony-stimulating factor, CSF-1) is a survival factor essential for the proliferation and development of monocytes, macrophages, and osteoclast progenitor cells. M-CSF also induces VEGF (vascular endothelial growth factor) secretion by macrophages, thereby mediating mobilization of endothelial progenitor cells and neovascularization. M-CSF is present as several bioactive isoforms that differ in potency and stability. The full-length protein is synthesized as a membrane-spanning protein that can be expressed on the cell surface or further cleaved and modified in the secretory vesicle. Further, M-CSF is a disulfide-bonded homodimer which is processed into one of two isoforms, a glycoprotein or a proteoglycan that has been modified by the addition of chondroitin sulfate to each subunit. Binding of M-CSF to its receptor, c-Fms (CSF-1R or CD115) induces dimerization of the receptor followed by internalization and degradation of the complex. Functionally, M-CSF is known to stimulate differentiation of hematopoietic stem cells to monocyte-macrophage cell populations in culture. M-CSF acts through the CSF receptor 1. Although human M-CSF shows activity on mouse cells, mouse CSF shows no activity on human cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07141
For Use With (Application) Control, Bioactivity
Format Lyophilized
Formulation Protein with no preservative
Gene ID (Entrez) 12977
Molecular Weight (g/mol) 36.4 kDa
Name Mouse M-CSF, Animal-Free
Purification Method Purified
Quantity 50 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias C87615; colony stimulating factor 1; colony stimulating factor 1 (macrophage); colony-stimulating factor-1 splice variant; CSF1; CSF-1; Csfm; H-MCSF; lanimostim; macrophage colony stimulating factor 1; macrophage colony stimulating factor alpha; macrophage colony stimulating factor beta; macrophage colony-stimulating factor; macrophage colony-stimulating factor 1; macrophage colony-stimulating factor beta; macrophage-colony stimulating factor alpha; MCSF; MCSF alpha; MCSFBETA; MGC31930; M-MCSF; op; osteopetrosis; Processed macrophage colony-stimulating factor 1
Common Name M-CSF
Gene Symbol Csf1
Biological Activity Determined by its ability to stimulate the proliferation of murine M-NFS-60 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKP
Content And Storage -20°C
Activity Determined by its ability to stimulate the proliferation of murine M-NFS-60 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.