Learn More
FUJIFILM BIOSCIENCES INC Mouse M-CSF, Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 2000810UG
Encompass_Preferred
Macrophage colony stimulating factor (M-CSF) is a hematopoietic growth factor that is widely produced by a variety of cells.  M-CSF stimulates the proliferation and differentiation of hematopoietic stem cells into monocyte and macrophage cell types.  M-CSF also acts through the colony stimulating factor 1 receptor (CSF1R) to modulate processes involved in immunology, bone metabolism, fertility, and pregnancy.  Human M-CSF shows activity on mouse cells; however, mouse M-CSF shows no activity on human cells.  
Catalog No. 50-990-816