Learn More
R&D Systems™ Mouse M-CSF Quantikine ELISA Kit

Specifications
Specifications
| Assay Range | 6.25 to 400 pg/mL |
| Assay Sensitivity | 0.486 pg/mL |
| Specificity | Natural and recombinant mouse M-CSF. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1435 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930 |
| Target | Mouse |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.