Learn More
ACROBiosystems Recombinant Protein;Mouse M-CSF R / CSF1R / CD115 Protein, Fc Tag, low endotoxin;HEK293;100UG;CSR,CSF1R,C-FMS,CD115,CSFR,FIM2,FMS,M-CSFR,CSF-1R,CSF1 R
Supplier:  ACROBiosystems CSRM5256100UG
Encompass_Preferred
Recombinant Protein;Mouse M-CSF R / CSF1R / CD115 Protein, Fc Tag, low endotoxin;100UG;Mouse M-CSF R, Fc Tag (CSR-M5256) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Ser 511 (Accession # NP_001032948).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 81.8 kDa. As a result of glycosylation, the protein migrates as 100-130 kDa under reducing (R) condition, and 260 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Mouse M-CSF Protein at 2μg/mL (100 μL/well) can bind Mouse M-CSF R, Fc Tag (Cat. No. CSR-M5256) with a linear range of 0.3-12.8 ng/mL (Routinely tested).
Catalog No. 50-113-8303