Learn More
American Research Products Inc Mouse Macrophage Colony Stimul
Catalog No. 5000329878
Encompass
Mouse Macrophage Colony-Stimulating Factor M-CSF ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29878 Supplier American Research Products Inc Supplier No. CSBE04659M
Mouse Macrophage Colony-Stimulating Factor M-CSF ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich