Learn More
American Research Products Inc Mouse Macrophage DeriVed Chemo
Catalog No. 5000326401
Encompass
Mouse Macrophage-DeriVed Chemokine MDC ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 23 672 pg/mL Sample serum plasma tissue homogenates cell culture supernates cell lysates Method Sandwich
Catalog No. 50-003-26401 Supplier American Research Products Inc Supplier No. CSBE04661M
Mouse Macrophage-DeriVed Chemokine MDC ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 23 672 pg/mL Sample serum plasma tissue homogenates cell culture supernates cell lysates Method Sandwich