Learn More
American Research Products Inc Mouse Macrophage metalloelasta
Catalog No. 5000328102
Encompass
Mouse Macrophage metalloelastase (MMP12) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates cell culture homogenates Method Sandwich
Catalog No. 50-003-28102 Supplier American Research Products Inc Supplier No. CSBEL014659MO
Mouse Macrophage metalloelastase (MMP12) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates cell culture homogenates Method Sandwich