Learn More
American Research Products Inc Mouse Macrophage Migration Inh
Catalog No. 5000329876
Encompass
Mouse Macrophage Migration Inhibitory Factor MIF ELISA Kit from CUSABIO Detection Range 125 pg/ml - 8000 pg/ml Sensitivity 31 25 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29876 Supplier American Research Products Inc Supplier No. CSBE07292M
Mouse Macrophage Migration Inhibitory Factor MIF ELISA Kit from CUSABIO Detection Range 125 pg/ml - 8000 pg/ml Sensitivity 31 25 pg/ml Sample serum plasma tissue homogenates Method Sandwich