Learn More
Biomatik Corporation Mouse Macrophage Stimulating Protein (MSP) ELISA Kit
Supplier:  Biomatik Corporation EKU05766
Encompass_Preferred
Mouse Macrophage Stimulating Protein (MSP) ELISA Kit for detecting MSP in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.47pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26928. Gene ID: 15235. Target Synonyms: MST1; HGFL; NF15S2; Hepatocyte Growth Factor-Like Protein; Macrophage Stimulating 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5836