Learn More
Sino Biological Recombinant Mouse MAdCAM-1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 58002M08H1
Encompass_Preferred
A DNA sequence encoding the Mouse MADCAM1 (NP_038619) (Met1-Ser364) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2949