Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse MAGP-2/MFAP5 (NP_056591.1) VersaClone cDNA

Catalog No. RDC3740
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC3740 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC3740 Supplier R&D Systems™ Supplier No. RDC3740
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Gene Wnt-6
Accession Number NP_056591.1
Gene ID (Entrez) 7475
Gene Alias protein Wnt-6, wingless-type MMTV integration site family, member 6, Wnt6
Content And Storage Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.