Learn More
Sino Biological Anti-MANF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50954T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse MANF (Catalog#50954-M08H; NP_083379.2; Met1-Leu179). MANF specific IgG was purified by Mouse MANF affinity chromatography.
Catalog No. 50-241-2171