Learn More
American Research Products Inc Mouse Mannan binding lectin se
Catalog No. 5000327448
Encompass
Mouse Mannan-binding lectin serine protease 2 (MASP2) ELISA kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 2 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27448 Supplier American Research Products Inc Supplier No. CSBEL013510MO
Mouse Mannan-binding lectin serine protease 2 (MASP2) ELISA kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 2 pg/mL Sample serum plasma tissue homogenates Method Competitive