Learn More
BIOHIPPO Mouse MAP1LC3B Microsample
SDP
Catalog No. 5000315724
Encompass
Mouse MAP1LC3B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MAP1LC3B in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 055 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MAP1LC3B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MAP1LC3B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MAP1LC3B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15724 Supplier BIOHIPPO Supplier No. BHE15210622
Mouse MAP1LC3B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MAP1LC3B in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 055 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MAP1LC3B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MAP1LC3B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MAP1LC3B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color