Learn More
Sino Biological Anti-MEK1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50312RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse MEK1 / MAP2K1 / MKK1 (rM MEK1 / MAP2K1 / MKK1; Catalog#50312-M20B; P31938; Met1-Ile393). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8320