Learn More
Abcam Recombinant mouse MAP4K1/HPK1 protein (Active), 5UG
Supplier:  Abcam AB2686425UG
Encompass_Preferred
Recombinant mouse MAP4K1/HPK1 protein (Active) is a Mouse Fragment protein, in the 1 to 346 aa range, expressed in Baculovirus infected Sf9, with >90% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Hpk1, Map4k1, Mitogen-activated protein kinase kinase kinase kinase 1, Hematopoietic progenitor kinase, MAPK/ERK kinase kinase kinase 1, HPK, MEK kinase kinase 1, MEKKK 1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1925150